Learn More
Selleck Chemical LLC Urapidil HCl S2025-50mg
SDP
Supplier:  Selleck Chemical LLC S202550MG
Encompass_Preferred
Urapidil HCl is a hydrochloride salt form of urapidil which is 1-adrenoceptor antagonist and 5-HT1A receptor agonist with pIC50 of 6 13 and 6 4 respectively
Catalog No. 50-311-0347