Learn More
Medchemexpress LLC URAT1 inhibitor 3 | 2850331-30-3 | C14H8Cl2N2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15143250MG
Encompass_Preferred
URAT1 inhibitor 3 is a potent, orally active, selective URAT1 inhibitor with an IC50 value of 0.8 nM. It demonstrates urate-lowering efficacy and is suitable for research on gout and hyperuricemia.
  • Potent, orally active, and selective URAT1 inhibitor
  • Has an IC50 value of 0.8 nM for URAT1
  • Demonstrates urate-lowering efficacy
  • Shows low toxicity at high concentrations in HepG2 and HK2 cells
  • Does not cause hepatic or renal toxicities in mice at tested dosages
  • Useful in research for gout and hyperuricemia
Catalog No. 50-003-72186