Learn More
Sanford Distributing Co High Peel Strength Urethane D-50, 100/carton
Supplier:  Sanford Distributing Co 04022
Encompass_Preferred
Double Bubble High Peel Strength Green & Beige Package has a Shore Hardness D-50 and is a good combination of high peel and lap shear strengths. It is semi-flexible, will bond to rubber, leather, china & many plastics. For automotive repairs, bonding aftermarket replacement mirrors, etc., repair of shoes, ski boots, and other sports equipment, repair of conveyor belts.
Catalog No. NC9214752