Learn More
Medchemexpress LLC URB937 10mg | 1357160-72-5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11647710MG
Encompass_Preferred
URB937 is an orally active, peripherally restricted inhibitor of fatty acid amide hydrolase (FAAH) used in pharmacological research. It raises peripheral anandamide levels without appreciable brain FAAH inhibition and is supplied as a solid for research use only.
  • Peripherally restricted FAAH inhibitor suitable for in vivo and in vitro studies.
  • Potent activity with reported IC50 ≈ 26.8 nM against FAAH.
  • High purity for improved experimental reliability.
  • Solid form with recommended cold storage to maintain stability.
  • Available in small-mass packages convenient for preclinical research.
Catalog No. 50-002-27853