Learn More
Medchemexpress LLC Urea-13C-15N2 | 58069-83-3 | MFCD00145506 | 100.0% | 63.03 | 13CH4(15N)2O | 1mg
SDP
Supplier:  Medchemexpress LLC HYY0271S21MG
Encompass_Preferred
Urea-13C 15N2 is the 13C and 15N labeled Urea[1] Urea is a powerful protein denaturant via both direct and indirect mechanisms[1] A potent emollient and keratolytic agent[2] Used as a diuretic agent Blood urea nitrogen (BUN) has been utilized to evaluate renal function[3] Widely used in fertilizers as a source of nitrogen and is an important raw material for the chemical industry
Catalog No. 50-002-67612