Learn More
Medchemexpress LLC Urea-d3, methyl-d3- | 56683-43-3 | 96.8% | 80.12 | C2D6N2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW015971S1MG
Encompass_Preferred
1-Methylureal-d6 is the deuterium-labeled (d6) isotopologue of 1-methylurea supplied as a stable isotope reference standard for analytical and research applications. It is used in mass spectrometry, method validation, and tracer studies, and a manufacturer certificate reports purity by HPLC.
  • Deuterium-labeled internal standard for mass spectrometry.
  • Useful for quantitative analysis and method validation.
  • Suitable for metabolic and tracer studies.
  • High purity confirmed by HPLC (96.8%).
  • Offered in mg-scale quantities for analytical use.
Catalog No. 50-002-99672