Learn More
Medchemexpress LLC Urea-d3, methyl-d3- | 56683-43-3 | MFCD06658635 | 96.8% | 80.12 g/mol | C2D6N2O | 5 MG

Supplier: Medchemexpress LLC HYW015971S5MG
1-Methylureal-d6 is the deuterium-labeled analogue of 1-methylurea intended for use as a stable isotope standard or tracer in analytical and research workflows. It is supplied as a solid with confirmed purity and defined molecular properties for reliable analytical performance.
- Deuterium-labeled compound suitable for use as a stable isotope standard or tracer.
- Purity 96.8% as confirmed by certificate of analysis.
- Solid, white to off-white appearance for easy handling.
- Molecular formula C2D6N2O and molecular weight 80.12 g/mol for accurate mass spectrometry applications.
- Recommended storage: powder at -20°C to maintain stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.