Learn More
Medchemexpress LLC Urea, N-[4-[4-(4-morpholinyl)-6-(3-oxa-8-azabicyclo[3.2.1]oct-8-yl)-1,3,5-triazin-2-yl]phenyl]-N'-4-pyridinyl-, hydrochloride (1:1) | 1463510-35-1 | 99.66% | 525.00 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY11080A10MG
Encompass_Preferred
PKI-179 hydrochloride is a potent and orally active dual PI3K/mTOR inhibitor. It exhibits anti-tumor activity in vivo, inhibiting tumor growth in nude mice bearing MDA-361 human breast cancer tumors. It also demonstrates good oral bioavailability and a high half-life.
  • Potent and orally active dual PI3K/mTOR inhibitor
  • Exhibits anti-tumor activity in vivo
  • Inhibits cell proliferation with low IC50s
  • Good oral bioavailability and high half-life
  • Well tolerated in animal models
Catalog No. 50-003-43743