Learn More
Medchemexpress LLC Urea, N-(4-chloro-2-hydroxy-3-sulfonylphenyl)-N'-(1R-2-methyl-2-cyclopenten-1-yl) | 1838123-22-0 | 99.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY120878A5MG
Encompass_Preferred
(R,R)-CXCR2-IN-2 is the (R,R) diastereomer of CXCR2-IN-2, a selective, brain-penetrant CXCR2 antagonist supplied for preclinical research. It is provided as solid samples and as a 10 mM solution in DMSO.
  • Selective CXCR2 antagonist suitable for pharmacology studies.
  • Brain-penetrant profile supports CNS-focused experiments.
  • High purity and high enantiomeric excess.
  • Available as solid samples and as a 10 mM solution in DMSO.
  • Molecular weight 414.90 g·mol⁻¹; formula C18H23ClN2O5S.
Catalog No. 50-002-31659