Learn More
Medchemexpress LLC Urea, N-(4-ethynylphenyl)-N'-1H-indol-3-yl- | 2244892-03-1 | 95.08% | 275.30 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17656650MG
Encompass_Preferred
H-151 Alkyne is a selective STING (Stimulator of interferon genes) inhibitor. It is a promising compound for research into autoimmune diseases such as systemic lupus erythematosus, scleroderma, and autoinflammatory diseases.
  • Selective STING inhibitor
  • Promising for autoimmune disease research
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-003-87436