Learn More
Medchemexpress LLC Urea, N,N'-bis(4-aminophenyl)- | 4550-72-5 | 99.9% | 242.28 | 250 MG

Supplier: Medchemexpress LLC 250MGHY108937
NSC 15364 is an inhibitor of VDAC1 oligomerization and apoptosis, intended for research use only and not for sale to patients.
- Solid appearance
- Gray to brown color
- Ships at room temperature in continental US; may vary elsewhere
- Storage for powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO: 50 mg/mL (206.37 mM); requires ultrasonic; hygroscopic DMSO has a significant impact on solubility, so newly opened DMSO should be used.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.