Learn More
Strem, An Ascensus Company CAS 1198080-53-3. 250mg. 1-(2R)-1-[(11bR)-Dinaphtho[21-d12-f][132]dioxaphosphepin-4-yloxy]propan-2-yl-3-phenylurea min. 97% UREAPhos. MFCD18827627
SDP
Supplier:  Strem, An Ascensus Company 152200250MG
Encompass
CAS 1198080-53-3. 250mg. 1-(2R)-1-[(11bR)-Dinaphtho[21-d12-f][132]dioxaphosphepin-4-yloxy]propan-2-yl-3-phenylurea min. 97% UREAPhos. MFCD18827627. Molecular Weight 508.50. Molecular Formula C30H25N2O4P. Color/form white pwdr. Strem 15-2200. http//www.strem.com/catalog/v/15-2200/
Catalog No. 50-211-0759