Learn More
ATCC Ureaplasma parvum
Supplier:  ATCC 700970
Encompass
This strain of Ureaplasma parvum is genome sequenced (GenBank AF222894). This bacterium is propagated in B-broth medium for Ureaplasma.
Catalog No. 50-238-5031