Learn More
Sigma Aldrich Fine Chemicals Biosciences Urease from Canavalia ensi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences U4002100KU
Encompass
Ureasenbsp(Ure) enzyme is present in the cytosol. It is active at acidic pH and is expressed at 28C. Urease binds to nickel and is present in archaea bacteria unicellular eukaryotes and plantsJack bean urease was the first enzyme to be crystallized and the first enzyme found to contain nickel. It is a multi-subunit enzyme consisting of 91 kDa subunits in three protein forms.
Catalog No. 50-290-3176