Learn More
CPC Scientific H-Asp-Leu-Trp-Gln-Lys-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific IMMO004B
Encompass
U5-peptide, a pentapeptide originally isolated from the ultrafiltrate of a uremic patient, shows immunomodulating properties.SEQUENCE: H-Asp-Leu-Trp-Gln-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: DLWQKMOLECULAR FORMULA: C32H48N8O9MOLECULAR WEIGHT: 688.78STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [71494-20-7 ]SYNONYMS: RESEARCH AREA: ImmunomodulationREFERENCES:D.Niese et al., Klin. Wschr., 64, 642 (1986)SKU(s): IMMO-004A, IMMO-004B
Catalog No. 50-210-9601