Learn More
Pharmacal Research Laboratories URID SCALE STRIP/DETERG/5GAL
SDP
Supplier:  Pharmacal Research Laboratories 83025F
Encompass
Detergent; Acid; Urid;; Strips urine scale quickly and efficiently from cages and trays, floors and walls; Penetrates, dissolves and releases the mineral bonding agents that cause adherent scale; 1.8 pH; Pink Liquid; 5 gal.
Catalog No. NC9045063