Learn More
Cayman Chemical UrIdIn-3-monphosphatesod 25mg
Supplier: Cayman Chemical 1677425 MG
A nucleoside used in the biochemical synthesis of ribothymidine-3’-phosphate that has been used as a bioavailable source of uridine in research studies designed to enhance learning and memory