Learn More
Cayman Chemical UrIdIn5O 3thIotrIphosph 2.5mg
SDP
Supplier:  Cayman Chemical 386922.5 MG
Encompass_Preferred
A P2Y2 receptor agonist; induces inositol phosphate formation in 1321N1 astrocytoma cells expressing the human P2Y2 receptor (EC50 = 240 nM) and chloride secretion in primary nasal epithelial cells from patients with cystic fibrosis in a concentration-dependent manner; induces vasoconstriction in isolated human coronary arteries (EC50 = 25.1 µM)
Catalog No. 50-291-9121