Learn More
Medchemexpress LLC Uridine (Standard) | 58-96-8 | 100.0% | 100 G

Supplier: Medchemexpress LLC HYB1449100G
Uridine (β-Uridine) is a nucleoside compound composed of uracil and a ribose ring, which are connected by a β-N1-glycosidic bond. It is for research use only.
- Nucleoside compound.
- High purity: 99.97%.
- Inhibits the proliferation of leukemia cell HL-60 and induces differentiation in vitro.
- Alleviates CCl4-induced liver fibrosis and inhibits pro-inflammatory cytokines and NF-κB signaling pathway activation in mouse models.
- Exhibits anti-ischemic, antioxidant, and anti-arrhythmic effects in rat models of acute myocardial ischemia and ischemia/reperfusion injury.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.