Learn More
Medchemexpress LLC Uridine-13C | 201996-62-5 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1449S11MG
Encompass_Preferred
Uridine-13C is the 13C labeled Uridine. It is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during the drug development process.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-004-12687