Learn More
Medchemexpress LLC Uridine 3',5'-diphosphate | 2922-95-4 | C9H14N2O12P2 | 100 MG

Supplier: Medchemexpress LLC HY154828100MG
Uridine 3',5'-diphosphate (3',5'-UDP) is a competitive RNase inhibitor. This product is a solid, white to off-white in color, with a high purity of 99.65%. It is soluble in H2O at 125 mg/mL (309.28 mM).
- Competitive RNase inhibitor
- Solid appearance
- White to off-white color
- Purity of 99.65%
- Soluble in H2O at 125 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.