Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 3′,5′-diphosphate | 2922-95-4 | C9H14N2O12P2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY15482850MG

Encompass_Preferred

Uridine 3′,5′-diphosphate (3′,5′-UDP), also referred to as Compound pUp, is a high-purity competitive RNase inhibitor. This chemical compound is presented as a white to off-white solid.

  • Suitable for research use only
  • Optimal storage for powder is -20°C for 3 years
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in H2O at 125 mg/mL (ultrasonic assistance may be needed)
  • Shipping handled at room temperature in continental US

Catalog No. 50-003-76052


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.