Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Uridine 3′,5′-diphosphate | 2922-95-4 | C9H14N2O12P2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1548285MG

Encompass_Preferred

Uridine 3′,5′-diphosphate is a competitive RNase inhibitor. It is for research use only and not sold to patients.

  • Purity: 99.65%
  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 404.16
  • Solubility in H2O: 125 mg/mL (needs ultrasonic)

Catalog No. 50-003-76418


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.