Learn More
Medchemexpress LLC Uridine 5'-(trihydrogen diphosphate), sodium salt (1:x) | 21931-53-3 | 98.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010820
Encompass_Preferred
Uridine 5'-diphosphate sodium salt is an endogenous metabolite that acts as a potent and selective native agonist for the P2Y6 receptor (EC50=300 nM) and a potent antagonist for the P2Y14 receptor. It catalyzes the glucuronidation of various substrates and is crucial for nucleic acid (RNA) biosynthesis. For research use only.
  • Potent and selective P2Y6 receptor agonist
  • Potent P2Y14 receptor antagonist
  • Catalyzes glucuronidation of various substrates
  • Used in nucleic acid (RNA) biosynthesis
  • Induces microglial CCL2 and CCL3 mRNA expression
  • Activates NFATc1 and NFATc2 in microglia
Catalog No. 50-004-54074