Learn More
Medchemexpress LLC Uridine-5'-diphosphoglucose pyrophosphorylase | 9026-22-6 | 200 U
SDP
Supplier:  Medchemexpress LLC HYP3018200U
Encompass_Preferred
Uridine-5'-diphosphoglucose pyrophosphorylase is involved in the catalysis of UDP-glucose synthesis. It can also be utilized to determine the concentration of pyrophosphate in urine.
  • Intended for research purposes
  • Available in various sizes
  • In-stock for immediate purchase
Catalog No. 50-003-98432