Learn More
Medchemexpress LLC Uridine, 5-ethynyl- | 69075-42-9 | MFCD00876719 | 99.2% | 268.22 | C11H12N2O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1411405MG
Encompass_Preferred
5-Ethynyluridine is a cell-permeable uridine analog used to label newly synthesized RNA in cell and in vivo models. It contains an alkyne functional group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC), allowing click-chemistry conjugation for isolation, sequencing, and visualization of nascent RNA.
  • Cell-permeable uridine analog for nascent RNA labeling.
  • Enables click-chemistry conjugation via alkyne (CuAAC).
  • Suitable for RNA isolation, sequencing, and imaging workflows.
  • Available as solid or ready-to-use DMSO solution formats.
  • High purity suitable for molecular biology applications.
  • Applicable in neuronal and in vivo transcription studies.
Catalog No. 50-002-23164