Learn More
Abcam Uridine-5'-monophosphate [UMP] disodium salt, nucleotide component of ribonucleic acid, 5G
Supplier:  Abcam AB1462235G
Encompass_Preferred
MW 368.15 g/mol, Purity >98%. Nucleotide that is a major component of ribonucleic acid. It is found in dietary supplements as well as natural RNA rich foods and has been shown to increase cognitive function in animals.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4828