Learn More
Medchemexpress LLC β-Uridine (Standard) | 58-96-8 | MFCD00006526 | 99.90% | 50 G

Supplier: Medchemexpress LLC HYB144950G
Uridine (β-Uridine) is a nucleoside compound composed of uracil and a ribose ring, which are connected by a β-N1-glycosidic bond. It has a molecular formula of C9H12N2O6 and a molecular weight of 244.20. It appears as a white to off-white solid. This product is for research use only and has not been fully validated for medical applications.
- A nucleoside compound
- Composed of uracil and a ribose ring
- Connected by a β-N1-glycosidic bond
- Molecular formula: C9H12N2O6
- Molecular weight: 244.20
- Appearance: white to off-white solid
- Storage: store at room temperature for 3 years
- In solvent, store at -80°C for 2 years or -20°C for 1 year
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.