Learn More
Medchemexpress LLC Uridine-d2-1 | 40632-21-1 | 98.5% | 5 MG
Supplier:  Medchemexpress LLC HYB1449S85MG
Uridine-d2-1 is the deuterium labeled Uridine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled uridine
  • Used as a tracer for quantitation during drug development
  • Can be used as an internal standard for quantitative analysis
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-13528