Learn More
Medchemexpress LLC Urocanic acid | 104-98-3 | 99.9% | 138.13 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1130081G
Encompass_Preferred
Urocanic acid is an endogenous metabolite found in the upper layers of mammalian skin. It serves as a significant absorber of ultraviolet radiation (UVR), playing a role in the skin's natural defense mechanisms. In vitro studies indicate its formation through the breakdown of filaggrin by proteinases.
  • Produced in upper layers of mammalian skin
  • Acts as a major absorber of ultraviolet radiation (UVR)
  • Formed from filaggrin breakdown in the epidermis
  • For research use only
Catalog No. 50-004-39736