Learn More
CPC Scientific H-Val-Ile-Leu-Ser-Leu-Asp-Val-Pro-Ile-Gly-Leu-Leu-Arg-Ile-Leu-Leu-Glu-Gln-Ala-Arg-Tyr-Lys-Ala-Ala-Arg-Asn-Gln-Ala-Ala-Thr-Asn-Ala-Gln-Ile-Leu-Ala-His-Val-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific UROC005A
Encompass
This peptide has exhibited considerably higher affinity for both splice variants of the type 2 CRF receptor (CRF-R2) than for those of CRF-R1. The potencies of Ucn II (mouse) in activating CRF-R2a and CRF-R2b were almost the same to that of urocortin (rat).ONE-LETTER SEQUENCE: VILSLDVPIGLLRILLEQARYKAARNQAATNAQILAHV-NH2MOLECULAR FORMULA: C187H320N56O50MOLECULAR WEIGHT:4153STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [330648-32-3], RESEARCH AREA: NeuropeptidesREFERENCES: K.Lewis et al., Proc. Natl. Acad. Sci. USA , 98, 7570 (2001); S.Y.Hsu et al., Nature Medicine 7, 605 (2001); T.M.Reyes et al., Proc. Natl. Acad. Sci. USA , 98, 2843 (2001)
Catalog No. 50-211-0594