Learn More
Selleck Chemical LLC Urolithin A S5312-100mg
SDP
Supplier:  Selleck Chemical LLC S5312100MG
Encompass_Preferred
Urolithin A (3 8-Dihydroxy Urolithin 2 7-Dihydroxy-3 4-benzocoumarin) a metabolite of ellagitannin is a first-in-class natural compound that induces mitophagy both in vitro and in vivo following oral consumption
Catalog No. 50-311-1724