Learn More
Medchemexpress LLC HY-135897 10mg Medchemexpress, Urolithin C CAS:165393-06-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13589710MG
Encompass_Preferred
Medchemexpress, HY-135897 10mg Urolithin C CAS:165393-06-6 Urolithin C, a gut-microbial metabolite of Ellagic acid, is a glucose-dependent activator of insulin secretion. Urolithin C is a L-type Ca2+ channel opener and enhances Ca2+ influx. Urolithin C induces cell apoptosis through a mitochondria-mediated pathway and also stimulates reactive oxygen species (ROS) formation[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0380