Learn More
Medchemexpress LLC Urotensin II (114-124), human (TFA) | 251293-28-4 | 99.8% | 1502.59 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1164A10MG
Encompass_Preferred
Urotensin II (114-124), human, is an 11-amino acid residue peptide that acts as a potent vasoconstrictor and agonist for the orphan receptor GPR14. It is intended for research use only.
  • 11-amino acid residue peptide.
  • Potent vasoconstrictor.
  • Agonist for the orphan receptor GPR14.
  • Binds to recombinant human GPR14 with high affinity, functionally coupled to calcium mobilization.
  • Induces concentration-dependent increases in intracellular calcium in HEK-293 cells expressing human GPR14 (EC50=0:62±0.17 nM).
  • Markedly increases total peripheral resistance in anesthetized non-human primates, associated with cardiac contractile dysfunction.
Catalog No. 50-004-22300