Learn More
Medchemexpress LLC Ursocholic acid | 2955-27-3 | 99.9% | 408.57 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113212100MG
Encompass_Preferred
Ursocholic acid is a bile acid found in mammalian bile that converts to deoxycholic acid (UDC) by mouse intestinal flora. It acts as a gallstone dissolving agent in the liver through anti-apoptosis, anti-inflammatory, immunomodulatory, bile regulation, and coordinated changes in mitochondrial integrity and cell signaling. It also shows favorable effects on bones in patients with chronic cholestasis.
  • Acts as a gallstone dissolving agent
  • Supports bone health in patients with chronic cholestasis
  • Promotes osteoblast differentiation and mineralization
  • Reduces inflammation and cytokine levels
Catalog No. 50-004-36523