Learn More
Cayman Chemical URSOCHOLIC ACID 5MG
SDP
Supplier:  Cayman Chemical 298835 MG
Encompass_Preferred
A bile acid; dietary administration decreases gallbladder cholesterol, phospholipid, and bile acid levels and the occurrence of gallstones in male mice fed a lithogenic diet at 0.25%
Catalog No. 50-298-8508