Learn More
Cayman Chemical UrsocholIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 354191 MG
Encompass_Preferred
An internal standard for the quantification of ursocholic acid by GC- or LC-MS
Catalog No. 50-291-3951