Learn More
Cayman Chemical UrsocholIc AcId-d4 5mg
SDP
Supplier:  Cayman Chemical 354195 MG
Encompass_Preferred
An internal standard for the quantification of ursocholic acid by GC- or LC-MS
Catalog No. 50-290-9585