Learn More
Medchemexpress LLC Ursodeoxycholic acid | 128-13-2 | MFCD00003680 | 392.57 | 1 G
SDP
Supplier:  Medchemexpress LLC HY137711G
Encompass_Preferred
Ursodeoxycholic acid (Ursodeoxycholate) is a secondary bile acid formed from the transformation of (cheno)deoxycholic acid by intestinal bacteria. It is a key regulator of intestinal barrier integrity and essential for lipid metabolism. It acts as a signaling molecule by interacting with bile acid-activated receptors, including G-protein coupled bile acid receptor 5 (TGR5, GPCR19) and the farnesoid X receptor (FXR). It can be used for research on various hepatic and gastrointestinal diseases. It also reduces ACE2 expression and is beneficial for reducing SARS-CoV-2 infection. It is orally active.
  • Regulates intestinal barrier integrity
  • Essential for lipid metabolism
  • Acts as a signaling molecule
  • Interacts with bile acid receptors (TGR5, GPCR19, FXR)
  • Used for hepatic and gastrointestinal disease research
  • Reduces ACE2 expression
  • Beneficial for reducing SARS-CoV-2 infection
  • Orally active
Catalog No. 50-003-51188