Learn More
Cayman Chemical UrsodeoxycholIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 218921 MG
Encompass_Preferred
An internal standard for the quantification of ursodeoxycholic acid by GC- or LC-MS
Catalog No. 50-298-7507