Learn More
Indofine Chemical Ursolic Acid, 98%, 250 Mg
SDP
Supplier:  Indofine Chemical 06031 250 MG
Encompass_Preferred
URSOLIC ACID 250 MG, Natural Products, 3b-Hydroxyurs-12-en-28-oic acid, C30H48O3, MW: 456.71, 0.98, 77-52-1, MFCD00009621
Catalog No. 50-908-206