Learn More
Innovative Research Inc US Cow Artry Cartd Frsh Saline
Supplier:  Innovative Research Inc IGBOARCS
Encompass_Preferred
Innovative Grade US Origin Bovine Artery Carotid Fresh in Saline - Innovative Grade US Origin Bovine Artery Carotid from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. Bovine Carotid Artery (adult) measures approximately 6-8 inches in length. This product is available in Adult (typically processed from animals fewer than 30 months of age) or Fetal Bovine. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4585