Learn More
Innovative Research Inc US Cow Umbilical Cord Fresh
Supplier:  Innovative Research Inc IGBOCDF
Encompass_Preferred
Innovative Grade US Origin Bovine Umbilical Cord Fresh - Innovative Grade US Origin Bovine Umbilical Cord from Innovative Research is collected on a custom basis from US Origin abattoir animals located in the United States, ensuring that only the highest-quality material is provided. This product is available fresh (refrigerated and shipped on wet ice) or frozen (stored at -20 to -80C and shipped on dry ice).
Catalog No. 50-274-4583