Learn More
Ushio America, Inc. USH-102D 100W Short Arc Mercury Medical Lamp
Supplier:  Ushio America, Inc. 5000273
Encompass_Preferred
The USHIO USH-102D, Hg 100W is a 100 watt, 20 volt clear glass mercury short arc lamp. This lamp has a 0.5 millimeter arc gap, a light center length of 43 millimeters and a luminous flux of 2200 lumens. Burn position for the USH-102D is vertical plus or minus 15 Deg.. USHIO's USH short-arc mercury lamps have a tightly confined plasma discharge, high luminance, and ultra-stable arc making them a good choice for microscopy units, diagnostic and industrial analytical equipment, blood analyzers, optical comparators, and UV curing applications.
Catalog No. NC0075362