Learn More
Bulbman Inc USHIO G25T8 GERMICIDAL T8
SDP
Supplier:  Bulbman Inc 122430
Encompass_Preferred
Ushio G25T8: Ushio Germicidal lamps are Low-Pressure Mercury-Arc lamps that emit radiation peaking at 253.7nm(UV-C). This output at 253.7nm is highly effective to inactivate microorganisms such as bacteria, viruses, yeast, and mold. Designed by experienced engineers, and by using quality material in combination with a tightly controlled manufacturing process, this high quality lamp is free of impurities to maintain strong and stable output throughout the life of the lamp. Ushio germicidal lamps are manufactured at an ISO9001 certified facility.
Catalog No. 50-125-3546