Learn More
U.S. Pharmacopeia Camphor, 464-49-3, MFCD00064149, 1 g
Supplier:  U.S. Pharmacopeia 1087508
Encompass
Empirical Formula (Hill Notation): C10H16O, Molecular Weight: 152.23, mp: 178-182 °C, Synonym: 2-Bornanone, 2-Camphanone, D-Camphor.
Catalog No. NC1447899