Learn More
U.S. Pharmacopeia Niacin, 59-67-6, MFCD00006391, 200mg
Supplier:  U.S. Pharmacopeia 1461003
Encompass
Molecular formula C6-H5-N-O2, Molecular weight 123.11 g/mol, Melting point 455 - 458.6 °F (235 - 237 °C), Flash point 379.40 °F (193.00 °C) (TOC), Synonyms: Nicotinic acid, 3-Pyridinecarboxylic acid
Catalog No. NC9458987