Learn More
Aviva Systems Biology Corp USP1 Antibody - C-terminal region (ARP59299_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59299_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against USP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human USP1. Alternate names: UBPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-651