Learn More
ABclonal Technology USP37 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15593500UL
Encompass_Preferred
Enables cysteine-type endopeptidase activity, protein kinase binding activity, and thiol-dependent deubiquitinase. Involved in G1/S transition of mitotic cell cycle, protein deubiquitination, and regulation of DNA replication. Located in nucleus.
Catalog No. 50-001-07271