Learn More
ABclonal Technology USP47 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15461500UL
Encompass_Preferred
Enables WD40-repeat domain binding activity and thiol-dependent deubiquitinase. Involved in several processes, including monoubiquitinated protein deubiquitination, negative regulation of G2/M transition of mitotic cell cycle, and negative regulation of nitrogen compound metabolic process. Located in cytoplasm. Part of SCF ubiquitin ligase complex.
Catalog No. 50-001-07317